siRNA Details
| Gene Symbol: Usp9y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148943.2 | NP_683745.2 | 11 | 1533 | ||
| GenBank mRNA | AJ307017.1 | - | 11 | 1533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp9y| Gene Symbol: Usp9y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148943.2 | NP_683745.2 | 11 | 1533 | ||
| GenBank mRNA | AJ307017.1 | - | 11 | 1533 | ||