siRNA Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148932.2 | NP_683734.2 | 9 | 1557 | ||
| GenBank mRNA | AF516680.1 | - | 9 | 1518 | ||
| AK154473.1 | - | 9 | 1558 | |||
| AK172982.1 | - | 8 | 935 | |||
| BC053101.1 | - | 9 | 1533 | 6405486 | ||

Pom121
Genomic Map