siRNA Details
| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081363.2 | NP_001074832.2 | 12 | 3564 | ||
| GenBank mRNA | AF194970.1 | - | 1 | 1849 | ||
| DQ642022.1 | - | 11 | 3421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpf| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081363.2 | NP_001074832.2 | 12 | 3564 | ||
| GenBank mRNA | AF194970.1 | - | 1 | 1849 | ||
| DQ642022.1 | - | 11 | 3421 | |||