siRNA Details
| Gene Symbol: Slc14a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171010.1 | NP_001164481.1 | 7 | 1132 | ||
| NM_001171011.1 | NP_001164482.1 | 6 | 867 | |||
| NM_028122.4 | NP_082398.1 | 6 | 927 | |||
| GenBank mRNA | AF448798.1 | - | 5 | 724 | ||
| AJ420967.1 | - | 6 | 786 | |||
| AK012066.1 | - | 6 | 777 | |||
| AK041979.1 | - | 6 | 869 | |||
| AK153891.1 | - | 7 | 1037 | |||
| BC058594.1 | - | 6 | 814 | 5364683 | ||
| BC086673.1 | - | 6 | 823 | 30646772 | ||
| BC100570.1 | - | 6 | 814 | 30044353 | ||

Slc14a1
Genomic Map