siRNA Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081977.2 | NP_001075446.1 | 6 | 699 | ||
| NM_080563.4 | NP_542130.1 | 5 | 625 | |||
| GenBank mRNA | AF360998.1 | - | 6 | 695 | ||
| AK133544.1 | - | 3 | 239 | |||
| AK172904.1 | - | 4 | 460 | |||
| BC030187.1 | - | 5 | 589 | 5345434 | ||
| BQ884505.1 | - | 5 | 558 | |||
| BY753828.1 | - | 5 | 618 | |||

Rnf144a
Genomic Map