siRNA Details
| Gene Symbol: Slco1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267707.1 | NP_001254636.1 | 9 | 1168 | ||
| NM_130861.3 | NP_570931.1 | 9 | 1116 | |||
| GenBank mRNA | AF240694.1 | - | 9 | 1086 | ||
| AK029706.1 | - | 9 | 1117 | |||
| AK166446.1 | - | 9 | 1145 | |||
| BC004814.1 | - | 2 | 225 | |||
| BC013594.1 | - | 9 | 1130 | 3586426 | ||

Slco1a5
Genomic Map