siRNA Details
| Gene Symbol: Fermt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153795.2 | NP_722490.1 | 10 | 1326 | ||
| GenBank mRNA | AK090332.1 | - | 10 | 1223 | ||
| AK142912.1 | - | 10 | 1327 | |||
| AK149737.1 | - | 10 | 1418 | |||
| AK158357.1 | - | 10 | 1223 | |||
| AK169682.1 | - | 10 | 1237 | |||
| BC025119.1 | - | 10 | 1258 | 4190760 | ||
| BC032204.1 | - | 10 | 1302 | 5342463 | ||
| BC066803.1 | - | 10 | 1287 | 30043017 | ||

Fermt3
Genomic Map