siRNA Details
| Gene Symbol: Slco4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148933.1 | NP_683735.1 | 2 | 779 | ||
| GenBank mRNA | AK033598.1 | - | 2 | 599 | ||
| AK172688.1 | - | 2 | 599 | |||
| BC030719.1 | - | 2 | 779 | 4501966 | ||
| BC030720.1 | - | 2 | 728 | 4505508 | ||

Slco4a1
Genomic Map