siRNA Details
| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081020.1 | NP_001074489.1 | 21 | 3017 | ||
| GenBank mRNA | AK030315.1 | - | 21 | 3955 | ||
| AK034499.1 | - | 4 | 402 | |||
| AK041499.1 | - | 2 | 233 | |||
| AK044528.1 | - | 21 | 3785 | |||
| BC172116.1 | - | 23 | 3111 | |||

Adamts6
Genomic Map