siRNA Details
| Gene Symbol: Rnpepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181405.4 | NP_852070.3 | 11 | 2045 | ||
| GenBank mRNA | AK075626.1 | - | 7 | 1311 | ||
| AK154881.1 | - | 8 | 3365 | |||
| BC024079.1 | - | 5 | 558 | |||
| BC031789.1 | - | 11 | 1465 | 4006916 | ||
| CA750931.1 | - | 3 | 412 | |||

Rnpepl1
Genomic Map