siRNA Details
| Gene Symbol: Ccdc86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023731.3 | NP_076220.2 | 4 | 1247 | ||
| GenBank mRNA | AB041663.1 | - | 4 | 1216 | ||
| AK016771.1 | - | 4 | 623 | |||
| AK036812.1 | - | 4 | 1238 | |||
| AK078407.1 | - | 4 | 1248 | |||
| AK078455.1 | - | 4 | 1248 | |||
| AK088727.1 | - | 4 | 1240 | |||
| BC043326.1 | - | 4 | 1243 | 5374087 | ||
| DQ501252.1 | - | 4 | 1214 | |||

Ccdc86
Genomic Map