siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133805.3 | NP_598566.3 | 2, 3 | 274 | ||
| GenBank mRNA | AF482000.1 | - | 2, 3 | 735 | ||
| AK079111.1 | - | 2, 3 | 208 | |||
| AK079522.1 | - | 2, 3 | 222 | |||
| AK088323.1 | - | 2, 3 | 194 | |||
| AK090013.1 | - | 2, 3 | 203 | |||
| AK145899.1 | - | 2, 3 | 201 | |||
| AK162502.1 | - | 2, 3 | 209 | |||
| AK195692.1 | - | 1, 2 | 49 | |||
| BC017690.1 | - | 2, 3 | 242 | 4238947 | ||
| BC021488.1 | - | 2, 3 | 233 | 5344864 | ||
| BC024421.1 | - | 2, 3 | 227 | 4987584 | ||

Cops8
Genomic Map