siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133805.3 | NP_598566.3 | 6 | 612 | ||
| GenBank mRNA | AF482000.1 | - | 6 | 1073 | ||
| AK079111.1 | - | 6 | 546 | |||
| AK079522.1 | - | 6 | 560 | |||
| AK088323.1 | - | 6 | 532 | |||
| AK090013.1 | - | 6 | 541 | |||
| AK142989.1 | - | 5 | 1581 | |||
| AK145899.1 | - | 6 | 539 | |||
| AK162502.1 | - | 6 | 547 | |||
| AK195692.1 | - | 5 | 387 | |||
| BC017690.1 | - | 6 | 580 | 4238947 | ||
| BC021488.1 | - | 6 | 571 | 5344864 | ||
| BC024421.1 | - | 6 | 565 | 4987584 | ||

Cops8
Genomic Map