siRNA Details
| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175367.6 | NP_780576.1 | 5 | 2271 | ||
| GenBank mRNA | AK036612.1 | - | 5 | 2272 | ||
| BC132531.1 | - | 4 | 1857 | 40130808 | ||
| BC138054.1 | - | 4 | 1857 | 8861073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ston2| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175367.6 | NP_780576.1 | 5 | 2271 | ||
| GenBank mRNA | AK036612.1 | - | 5 | 2272 | ||
| BC132531.1 | - | 4 | 1857 | 40130808 | ||
| BC138054.1 | - | 4 | 1857 | 8861073 | ||