siRNA Details
| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175370.4 | NP_780579.1 | 9, 10 | 806 | ||
| GenBank mRNA | AK016678.1 | - | 8, 9 | 1853 | ||
| AK077190.1 | - | 9, 10 | 774 | |||
| AK133373.1 | - | 804 | ||||
| BC145039.1 | - | 8, 9 | 713 | |||
| BC145040.1 | - | 8, 9 | 711 | |||

Als2cr12
Genomic Map