siRNA Details
| Gene Symbol: Jmjd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242396.1 | NP_001229325.1 | 9 | 4930 | ||
| NM_207221.2 | NP_997104.2 | 10 | 4698 | |||
| GenBank mRNA | AK082106.1 | - | 1 | 823 | ||
| AK173162.1 | - | 9 | 4700 | |||
| BC054559.1 | - | 1 | 1261 | |||
| BC068318.1 | - | 1 | 1255 | |||

Jmjd1c
Genomic Map