siRNA Details
| Gene Symbol: Ibtk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081282.2 | NP_001074751.1 | 12 | 2742 | ||
| GenBank mRNA | AK030895.1 | - | 12 | 2705 | ||
| AK031796.1 | - | 1 | 543 | |||
| AK050176.1 | - | 12 | 2735 | |||
| AK129353.1 | - | 9 | 1620 | |||
| BC050255.1 | - | 1 | 47 | |||
| BC060722.1 | - | 11 | 2536 | |||
| BC137798.1 | - | 12 | 2631 | 8860817 | ||

Ibtk
Genomic Map