siRNA Details
| Gene Symbol: Rdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290409.1 | NP_001277338.1 | 6 | 968 | ||
| NM_001290411.1 | NP_001277340.1 | 7 | 1106 | |||
| NM_175372.4 | NP_780581.1 | 7 | 1073 | |||
| GenBank mRNA | AK028434.1 | - | 7 | 1066 | ||
| AK034180.1 | - | 913 | ||||
| BC082583.1 | - | 7 | 1025 | 30613493 | ||

Rdh13
Genomic Map