siRNA Details
| Gene Symbol: Rdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290409.1 | NP_001277338.1 | 3 | 470 | ||
| NM_001290411.1 | NP_001277340.1 | 3 | 503 | |||
| NM_175372.4 | NP_780581.1 | 3 | 470 | |||
| GenBank mRNA | AK028434.1 | - | 3 | 463 | ||
| AK034180.1 | - | 415 | ||||
| AK089458.1 | - | 3 | 876 | |||
| BC082583.1 | - | 3 | 422 | 30613493 | ||

Rdh13
Genomic Map