siRNA Details
| Gene Symbol: Trmt10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175389.4 | NP_780598.2 | 7 | 736 | ||
| GenBank mRNA | AK089967.1 | - | 7 | 831 | ||
| AK145681.1 | - | 6 | 3160 | |||
| AK146481.1 | - | 7 | 868 | |||
| BC022121.1 | - | 6 | 626 | |||
| BC090650.1 | - | 7 | 736 | 30651613 | ||
| BC098478.1 | - | 7 | 801 | 30033713 | ||
| BC141035.1 | - | 7 | 724 | 9055647 | ||

Trmt10a
Genomic Map