siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242407.1 | NP_001229336.1 | 15, 16 | 1795 | ||
| NM_175392.3 | NP_780601.1 | 15, 16 | 1963 | |||
| GenBank mRNA | AK077618.1 | - | 15, 16 | 1756 | ||
| AK131152.1 | - | 11, 12 | 3537 | |||
| AK154200.1 | - | 15, 16 | 1784 | |||
| AK165996.1 | - | 15, 16 | 1787 | |||
| BC038823.1 | - | 3, 4 | 205 | |||
| BC051404.1 | - | 15, 16 | 1963 | 6395030 | ||

Fam73b
Genomic Map