siRNA Details
| Gene Symbol: Tbc1d10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178650.3 | NP_848765.2 | 3, 4 | 295 | ||
| GenBank mRNA | AK040487.1 | - | 3, 4 | 296 | ||
| AK075803.1 | - | 3, 4 | 282 | |||
| AK088119.1 | - | 360 | ||||
| AK131177.1 | - | 2, 3 | 276 | |||
| AK153836.1 | - | 3, 4 | 293 | |||
| AK154444.1 | - | 2, 3 | 273 | |||
| BC094901.1 | - | 2, 3 | 270 | |||
| BQ829173.1 | - | 3, 4 | 531 | |||

Tbc1d10c
Genomic Map