siRNA Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004561.3 | NP_004552.2 | 4 | 1783 | ||
| XM_005274018.3 | XP_005274075.1 | 4 | 1601 | |||
| XM_011545067.2 | XP_011543369.1 | 4 | 1599 | |||
| XM_011545068.2 | XP_011543370.1 | 4 | 2144 | |||
| XM_017017837.1 | XP_016873326.1 | 4 | 2437 | |||
| XM_017017838.1 | XP_016873327.1 | 4 | 1476 | |||
| GenBank mRNA | AF016045.1 | - | 3 | 1659 | ||
| BC043146.1 | - | 4 | 1374 | |||
| BC059408.1 | - | 4 | 1759 | 30343789 | ||
| BX648725.1 | - | 4 | 1663 | |||

OVOL1
Genomic Map