siRNA Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175400.6 | NP_780609.4 | 7 | 1092 | ||
| GenBank mRNA | AK044971.1 | - | 7 | 1029 | ||
| AK047631.1 | - | 2 | 123 | |||
| AK053638.1 | - | 7 | 1127 | |||
| BC066037.1 | - | 7 | 1091 | 6405661 | ||

Sephs1
Genomic Map