siRNA Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175400.6 | NP_780609.4 | 5 | 796 | ||
| GenBank mRNA | AK044971.1 | - | 5 | 733 | ||
| AK053638.1 | - | 5 | 831 | |||
| BC065165.1 | - | 5 | 757 | 6417054 | ||
| BC066037.1 | - | 5 | 795 | 6405661 | ||

Sephs1
Genomic Map