siRNA Details
| Gene Symbol: Uhrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144873.2 | NP_659122.2 | 12 | 2147 | ||
| GenBank mRNA | AB116653.1 | - | 12 | 1882 | ||
| AF274047.1 | - | 12 | 2057 | |||
| AK075912.1 | - | 1 | 92 | |||
| AK170648.1 | - | 3 | 303 | |||
| BC060241.1 | - | 12 | 2147 | 6334856 | ||

Uhrf2
Genomic Map