siRNA Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175403.3 | NP_780612.2 | 4, 5 | 788 | ||
| GenBank mRNA | AK031525.1 | - | 4, 5 | 758 | ||
| AK075940.1 | - | 4, 5 | 772 | |||
| AK129066.1 | - | 4, 5 | 691 | |||
| BC035300.1 | - | 4, 5 | 753 | 4237864 | ||
| BC058245.1 | - | 4, 5 | 742 | 6313259 | ||

Mlec
Genomic Map