siRNA Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163505.1 | NP_001156977.1 | 13 | 1832 | ||
| NM_146091.4 | NP_666203.3 | 13 | 1734 | |||
| GenBank mRNA | AK028842.1 | - | 13 | 1595 | ||
| AK030660.1 | - | 13 | 1594 | |||
| AK077752.1 | - | 13 | 1597 | |||
| AK147720.1 | - | 13 | 1692 | |||
| AK169476.1 | - | 13 | 1594 | |||
| AK170550.1 | - | 13 | 3109 | |||
| BC002149.1 | - | 10 | 1124 | |||
| BC017138.1 | - | 13 | 1583 | 4485115 | ||

Atl3
Genomic Map