siRNA Details
| Gene Symbol: Sobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175407.3 | NP_780616.4 | 2 | 578 | ||
| GenBank mRNA | BC057055.1 | - | 2 | 257 | ||
| BC059851.1 | - | 2 | 578 | 5704623 | ||
| DQ157775.1 | - | 2 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sobp| Gene Symbol: Sobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175407.3 | NP_780616.4 | 2 | 578 | ||
| GenBank mRNA | BC057055.1 | - | 2 | 257 | ||
| BC059851.1 | - | 2 | 578 | 5704623 | ||
| DQ157775.1 | - | 2 | 137 | |||