siRNA Details
| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027321.3 | NP_081597.1 | 5 | 639 | ||
| NM_175413.3 | NP_780622.1 | 5 | 639 | |||
| GenBank mRNA | AK008119.1 | - | 5 | 607 | ||
| AK035072.1 | - | 5 | 641 | |||
| AK039277.1 | - | 5 | 636 | |||
| AK046671.1 | - | 5 | 535 | |||
| AK052159.1 | - | 5 | 591 | |||
| AK079125.1 | - | 5 | 617 | |||
| BC119529.2 | - | 5 | 588 | 40056120 | ||
| BC119530.1 | - | 5 | 589 | 40056121 | ||

Lrrc39
Genomic Map