siRNA Details
| Gene Symbol: Tspan9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175414.4 | NP_780623.1 | 7, 8 | 755 | ||
| GenBank mRNA | AK079139.1 | - | 7, 8 | 728 | ||
| AK158729.1 | - | 6, 7 | 640 | |||
| AK163395.1 | - | 7, 8 | 811 | |||
| AK203288.1 | - | 1, 2 | 101 | |||
| BC052503.1 | - | 7, 8 | 737 | 5146854 | ||

Tspan9
Genomic Map