siRNA Details
| Gene Symbol: Mybpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252372.1 | NP_001239301.1 | 3, 4 | 399 | ||
| NM_175418.5 | NP_780627.2 | 4, 5 | 441 | |||
| GenBank mRNA | AK132188.1 | - | 3, 4 | 273 | ||
| BC062158.1 | - | 4, 5 | 303 | 30134842 | ||
| HQ848554.1 | - | 6, 7 | 289 | |||

Mybpc1
Genomic Map