siRNA Details
| Gene Symbol: R3hdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177994.4 | NP_818775.2 | 7 | 720 | ||
| GenBank mRNA | AK081288.1 | - | 7 | 709 | ||
| AK088107.1 | - | 7 | 721 | |||
| AK139047.1 | - | 7 | 711 | |||
| AK159869.1 | - | 7 | 698 | |||
| AK165610.1 | - | 7 | 691 | |||
| BC034653.1 | - | 6 | 523 | |||
| BC040686.1 | - | 4 | 352 | |||
| BC095970.1 | - | 7 | 701 | 6530790 | ||
| BC132463.1 | - | 7 | 796 | 40130740 | ||
| BC138461.1 | - | 7 | 796 | 8861481 | ||

R3hdm4
Genomic Map