siRNA Details
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 606 | ||
NM_001256522.1 | NP_001243451.1 | 2 | 446 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 606 | ||
NM_001256522.1 | NP_001243451.1 | 2 | 446 |