siRNA Details
| Gene Symbol: Rnf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163263.1 | NP_001156735.1 | 5 | 1084 | ||
| NM_182999.2 | NP_892044.1 | 6 | 931 | |||
| GenBank mRNA | AK043896.1 | - | 1 | 60 | ||
| AK048862.1 | - | 5 | 1043 | |||
| AK051278.1 | - | 5 | 1043 | |||
| AK053783.1 | - | 6 | 919 | |||
| AK082529.1 | - | 5 | 988 | |||
| AK087860.1 | - | 5 | 1012 | |||
| AK163905.1 | - | 2 | 297 | |||
| AK220492.1 | - | 5 | 1000 | |||
| BC004593.1 | - | 1 | 158 | |||
| BC052482.1 | - | 6 | 870 | 6308280 | ||

Rnf20
Genomic Map