siRNA Details
| Gene Symbol: Rnf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163263.1 | NP_001156735.1 | 12 | 1969 | ||
| NM_182999.2 | NP_892044.1 | 13 | 1816 | |||
| GenBank mRNA | AK048862.1 | - | 12 | 1928 | ||
| AK076501.1 | - | 8 | 898 | |||
| AK139883.1 | - | 10 | 1344 | |||
| AK163905.1 | - | 9 | 1181 | |||
| AK220492.1 | - | 12 | 1885 | |||
| BC052482.1 | - | 13 | 1755 | 6308280 | ||

Rnf20
Genomic Map