siRNA Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133974.3 | NP_598735.2 | 2 | 326 | ||
| GenBank mRNA | AK035674.1 | - | 2 | 288 | ||
| AK087108.1 | - | 2 | 224 | |||
| BC018410.1 | - | 2 | 337 | |||
| BC059937.1 | - | 2 | 233 | |||
| BC085253.1 | - | 2 | 326 | 5347122 | ||

Cdcp1
Genomic Map