siRNA Details
| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033227.2 | NP_001028399.1 | 11 | 1301 | ||
| GenBank mRNA | AK144033.1 | - | 11 | 1302 | ||
| AK165337.1 | - | 10 | 1097 | |||
| BC141325.1 | - | 10 | 1094 | 9055937 | ||

Slc5a10
Genomic Map