siRNA Details
| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033227.2 | NP_001028399.1 | 5 | 584 | ||
| GenBank mRNA | AK144033.1 | - | 5 | 585 | ||
| AK165337.1 | - | 4 | 380 | |||
| BC141325.1 | - | 4 | 377 | 9055937 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a10| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033227.2 | NP_001028399.1 | 5 | 584 | ||
| GenBank mRNA | AK144033.1 | - | 5 | 585 | ||
| AK165337.1 | - | 4 | 380 | |||
| BC141325.1 | - | 4 | 377 | 9055937 | ||