siRNA Details
| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033227.2 | NP_001028399.1 | 15 | 1908 | ||
| GenBank mRNA | AK082798.1 | - | 4 | 564 | ||
| AK144033.1 | - | 15 | 1909 | |||
| AK165337.1 | - | 14 | 1704 | |||
| BC141325.1 | - | 14 | 1701 | 9055937 | ||

Slc5a10
Genomic Map