siRNA Details
| Gene Symbol: Fam163b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175427.4 | NP_780636.1 | 3 | 534 | ||
| GenBank mRNA | AK083284.1 | - | 3 | 532 | ||
| AK162827.1 | - | 3 | 535 | |||
| BC147286.1 | - | 3 | 380 | 9056331 | ||
| BC147287.1 | - | 3 | 380 | 9056348 | ||

Fam163b
Genomic Map