siRNA Details
| Gene Symbol: Lmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207222.2 | NP_997105.1 | 3 | 253 | ||
| GenBank mRNA | AK034177.1 | - | 3 | 254 | ||
| BC057086.1 | - | 2 | 489 | 5720992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo3| Gene Symbol: Lmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207222.2 | NP_997105.1 | 3 | 253 | ||
| GenBank mRNA | AK034177.1 | - | 3 | 254 | ||
| BC057086.1 | - | 2 | 489 | 5720992 | ||