siRNA Details
| Gene Symbol: Lmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA050915.1 | - | 2 | 205 | ||
| AK133586.1 | - | 2 | 94 | |||
| AW494084.1 | - | 2 | 288 | |||
| BC053074.1 | - | 2 | 344 | 5693266 | ||
| DV662082.1 | - | 2 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo1| Gene Symbol: Lmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA050915.1 | - | 2 | 205 | ||
| AK133586.1 | - | 2 | 94 | |||
| AW494084.1 | - | 2 | 288 | |||
| BC053074.1 | - | 2 | 344 | 5693266 | ||
| DV662082.1 | - | 2 | 555 | |||