siRNA Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145575.3 | NP_663550.1 | 9, 10 | 1605 | ||
| GenBank mRNA | AK148009.1 | - | 7, 8 | 977 | ||
| BC019435.1 | - | 9, 10 | 1564 | 5148205 | ||
| CX568419.1 | - | 4, 5 | 301 | |||

Cald1
Genomic Map