siRNA Details
| Gene Symbol: Txlna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005506.3 | NP_001005506.2 | 5 | 1321 | ||
| NM_001199695.1 | NP_001186624.1 | 6 | 1038 | |||
| GenBank mRNA | AK135290.1 | - | 5 | 1169 | ||
| AK141252.1 | - | 6 | 972 | |||
| AK150240.1 | - | 6 | 995 | |||
| AK160829.1 | - | 5 | 1169 | |||
| AK161649.1 | - | 6 | 1002 | |||
| AK168418.1 | - | 5 | 1322 | |||
| AK171101.1 | - | 6 | 1014 | |||
| BC060227.1 | - | 5 | 1156 | 30357541 | ||
| BC065115.1 | - | 5 | 1156 | 30362350 | ||

Txlna
Genomic Map