siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177231.2 | NP_796205.1 | 8 | 902 | ||
| NM_178220.3 | NP_835738.1 | 8 | 902 | |||
| GenBank mRNA | AK052588.1 | - | 8 | 679 | ||
| AK090090.1 | - | 8 | 729 | |||
| AK144054.1 | - | 8 | 701 | |||
| AK147508.1 | - | 8 | 903 | |||
| AK147566.1 | - | 8 | 924 | |||
| AK165514.1 | - | 8 | 701 | |||
| BC108969.1 | - | 8 | 669 | 40047224 | ||
| BC108970.1 | - | 8 | 669 | 40047225 | ||

Arrb1
Genomic Map