siRNA Details
| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025350.3 | NP_079626.2 | 10 | 1370 | ||
| GenBank mRNA | AK003088.1 | - | 10 | 1154 | ||
| AK007330.1 | - | 4 | 439 | |||
| BC052661.1 | - | 10 | 1384 | 30050094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa1| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025350.3 | NP_079626.2 | 10 | 1370 | ||
| GenBank mRNA | AK003088.1 | - | 10 | 1154 | ||
| AK007330.1 | - | 4 | 439 | |||
| BC052661.1 | - | 10 | 1384 | 30050094 | ||