siRNA Details
| Gene Symbol: Itga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033228.3 | NP_001028400.2 | 13 | 1922 | ||
| GenBank mRNA | AK035870.1 | - | 13 | 1923 | ||
| AK040830.1 | - | 4 | 346 | |||
| AK043769.1 | - | 13 | 1861 | |||
| AK137169.1 | - | 13 | 1687 | |||

Itga1
Genomic Map