siRNA Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163746.1 | NP_001157218.1 | 2 | 229 | ||
| NM_028352.4 | NP_082628.3 | 2 | 229 | |||
| GenBank mRNA | AK013402.1 | - | 2 | 212 | ||
| AK028066.1 | - | 2 | 219 | |||
| AK032776.1 | - | 2 | 181 | |||
| AK036561.1 | - | 2 | 215 | |||
| AK043547.1 | - | 2 | 230 | |||
| AK049337.1 | - | 2 | 181 | |||
| AK049758.1 | - | 2 | 216 | |||
| AK051706.1 | - | 2 | 169 | |||
| AK153501.1 | - | 1 | 49 | |||
| AK160913.1 | - | 2 | 230 | |||
| AK165513.1 | - | 2 | 214 | |||
| AK169082.1 | - | 2 | 194 | |||
| AK169141.1 | - | 2 | 188 | |||
| AK169787.1 | - | 2 | 203 | |||
| BC138700.1 | - | 2 | 154 | 8861722 | ||

Pgm3
Genomic Map