siRNA Details
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243837.1 | NP_001230766.1 | 9 | 1115 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243837.1 | NP_001230766.1 | 9 | 1115 |